About this item
Reliably measure pH, conductivity and temperature with the Orion™ Star™ A215 pH/Conductivity Benchtop Meter for advanced laboratory analysis and interfacing.
- Combined convenience with the benefits of a pH meter and conductivity meter in one easy-to-use system
- Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
- Calibration editing option allows individual points to be fixed without a full recalibration
- Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
- Auto-Read mode holds a stable reading, Continuous mode shows changing readings with hold option, and Timed mode logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe to uniformly mix solutions
- Three-year meter warranty
Record accurate and reliable pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity, and temperature measurements with the Orion™ Star™ A215 pH/conductivity benchtop meter.
Orion™ Star™ A215 pH/Conductivity Benchtop Meters are easy to use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator provide flexibility and control over testing. All critical data is shown with measurements including temperature, date and time, read type, measurement stability, active calibration information, and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates. Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view, and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key to quickly access, evaluate, and change measurement and instrument settings. Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously. Flexible power options with universal power adapter or optional AA batteries. IP54-rated housing resists dust and splashes, and can be wall-mounted. Non-volatile meter memory ensures data and settings are protected, even with loss of power.
The unit is ideal for a wide range of applications and advanced pH and conductivity analysis in the laboratory. Use the two meter channels to measure pH and conductivity simultaneously or view each channel separately. Use linear, non-linear, EP (USP), or off conductivity temperature compensation with 5, 10, 15, 20, or 25°C reference temperatures. Perform up to a 5 point calibration, log up to 2000 data point sets with time/date stamp, and easily transfer calibration and data logs via USB or RS232 to a printer or computer.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89206-340
- 89206-342
- Resistivity resolution:
- 1 ohm or 0.1 meg-ohm, auto ranging
- 1 ohm or 0.1 meg-ohm, auto ranging
- Temperature resolution:
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- Temperature accuracy:
- ±0.1°C
- ±0.1°C
- Operating temperature range:
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- Memory:
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- No. of channels:
- 2
- 2
- Calibration:
- pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option and temperature
- pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option and temperature