About this item
Accurately monitor pH, mV, ORP, and temperature with the Orion™ Star™ A321 pH Portable Meter. The unit ensures waterproof, high performance in challenging field locations.
- Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
- Calibration editing option allows individual points to be fixed without a full recalibration
- Capture critical readings with measurement stability indicator and selectable read modes
- Auto-Read mode holds a stable reading, Continuous mode shows changing readings with hold option, and Timed mode logs data at specific intervals
- Calibration log stores ten most recent calibrations
- IP67-rated housing is waterproof for rugged field operation
- Three-year meter warranty
Get high accuracy and premium performance everywhere needed with the Orion™ Star™ A321 pH portable meter, which is designed for a wide range of pH, mV, ORP, and temperature testing and field applications.
Take measurements at the source with this premium line of portable meters. The Orion™ Star™ A321 pH Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements. These meters are designed to be easy to use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms, and password-protected measurement methods. Flexible power options with AA batteries or optional universal power adapter. Non-volatile meter memory ensures data and settings are protected, even with loss of power.
Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Perform up to a five point pH calibration, log up to 5000 data point sets with time/date stamp, and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion™ Star Com software.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89206-350
- 89206-348
- Temperature resolution:
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- Temperature accuracy:
- ±0.1°C
- ±0.1°C
- Battery life:
- 800 hours
- 800 hours
- Operating temperature range:
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- Memory:
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- No. of channels:
- 1
- 1
- Calibration:
- pH with calibration editing option, relative mV (RmV), ORP and temperature
- pH with calibration editing option, relative mV (RmV), ORP and temperature