About this item
CORIO™ refrigerated/heating circulators for routing liquid temperature control applications from –40 to +150 °C. A great value with German engineering and quality.
- Stainless steel construction
- Adjustable pump
- German quality
- USB communication
- Quiet operation
The new generation of economy CORIO™ refrigerated/heating circulators from JULABO provide fast and reliable temperature control for internal or external applications.
The units incorporate a USB port for connection to an external PC. This enables control via PLC or the free JULABO EasyTEMP software. The CORIO can also accommodate data–logging of time/temperature information with file downloading to a USB stick for data capture and analysis via Excel.
Units feature easy pump change–over for internal or external circulation. The units come with M16x1 male threaded pump connections with a pair of 8 and 12mm hose barbs. A space–saving cooling coil design yields more usable space in the bath tank. All systems utilize front to back air cooling with flat side panels. This minimizes the require laboratory installation space as the units can be placed next to a wall or other applications; only 20 cm of clearance in front and back are needed.
Specifications
- Catalog No:
- 76558-212
- 76558-216
- 76558-234
- 76558-166
- CA75871-446
- CA75839-566
- CA75839-424
- CA10015-750
- CA75839-422
- CA75839-420
- 77442-068
- 77442-070
- 77442-072
- 77442-074
- 76558-268
- 77442-060
- 76558-262
- 77442-062
- 76558-190
- 77442-064
- 76558-240
- 77442-066
- 76558-200
- 76558-272
- CA75871-450
- CA75871-448
- Environmentally Preferable:
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Water Efficient
- Pump:
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- 17 L/min; 6.2 psi max.
- 17 L/min; 6.2 psi max.
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- 16 L/min; 4.8 psi
- 16 L/min; 4.8 psi
- 16 L/min; 4.8 psi
- 16 L/min; 4.8 psi
- 17 L/min; 6.2 psi max.
- 15 L/min; 5.07 psi
- 15 L/min; 4.8 psi max.
- 15 L/min; 5.07 psi
- 15 L/min; 4.8 psi max.
- 16 L/min; 4.8 psi
- 15 L/min; 4.8 psi max.
- 16 L/min; 4.8 psi
- 15 L/min; 4.8 psi max.
- 17 L/min; 6.2 psi max.
- 15 L/min; 4.8 psi max.
- 15 L/min; 4.8 psi max.
- Stability:
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- ±0.03 °C
- Environmental certification:
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
- RoHS; WEEE
Specifications
Related Information
10015-750 data.pdf 10015-752 data.pdf 10015-754 data.pdf 10015-818 data.pdf JULABO-CORIO-CD-1000F--9012707.pdf
Datasheet
Julabo CORIO Immersion Circulator
Product Literature
Manual 1000FW Basic Refrigerated Bath Units.pdf Manual CORIO™ CD Heating Immersion Circulator Manual Heating immersion circulator, open bath heating circulator, refrigerated circulator 1.950.0800.us.V10.pdf Manual Refrigerated Bath Basic Units Manual Refrigerated bath basic units 1.950.0170.us.V07