UV Safety Goggles, Azure Biosystems
Catalog #: 10001-820
Supplier: Azure Biosystems
Product available on GSA Advantage®
Specifications
- Description:UV safety goggles
- Supplier No.:AC1022
Specifications
About this item
Safety goggle used while working with UV light or blue light.
Recommended to be used for band excision.
Customers Who Bought This Also Bought