About this item
Get premium performance and ultimate flexibility with the Orion™ Versa Star Pro™ pH Benchtop Meter for pH, mV, ORP, and temperature on customizable color display.
- Onscreen measurement stability indicator lets users know exactly when measurements are ready
- Auto-Read mode holds a stable reading, Continuous mode shows changing readings, Timed mode logs data at specific intervals, and Single-Shot mode holds a reading after a preset time is reached
- Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
- Easily access, update and switch preferred measurement settings, using ten password-protected methods per channel
- Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position and can be quickly rinsed between samples
- Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID, and account ID
- Three-year meter and module warranty
Get high accuracy, premium performance and ultimate flexibility with the Orion™ Versa Star Pro™ Benchtop Meter with pH Module.
Orion™ Versa Star Pro™ benchtop meters are designed to meet the most challenging applications with ease. Delivering rich functionality and features, users can optimally configure the meter system to meet changing requirements. These meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view measurements thanks to a color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad. Calibration log stores ten most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations. Ensure measurement integrity by viewing the active calibration log while taking readings. Meter facilitates data transfers to a computer via USB or RS232 connections. Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values. Calibration editing option allows individual points to be fixed without a full recalibration. Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading. Non-volatile meter memory preserves data and settings, even in the event of power loss. IP54-rated housing resists dust and splashes, and can be wall-mounted if table space is limited.
Meet the most challenging applications for pH, mV, ORP, and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Perform up to a six point pH calibration, log up to 2000 data point sets with time/date stamp, and easily transfer calibration and data logs via USB or RS232 to a printer or computer. Use the included electrode stand to easily place sensors in samples.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89206-380
- 89206-378
- 76076-768
- Temperature resolution:
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- 0.1°C, 0.1°F
- Temperature accuracy:
- ±0.1°C
- ±0.1°C
- ±0.1°C
- Operating temperature range:
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- 5…45 °C (41…113 °F)
- Memory:
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- Non-volatile memory preserves data log, calibration log, and meter settings
- No. of channels:
- 4 - Customizable with modules
- 4 - Customizable with modules
- 4 - Customizable with modules
- Input:
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 2 stirrer inputs (stirrer probe)
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 2 stirrer inputs (stirrer probe)
- BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 2 stirrer inputs (stirrer probe)
- Calibration:
- pH with calibration editing option, relative mV (RmV), ORP and temperature
- pH with calibration editing option, relative mV (RmV), ORP and temperature
- pH with calibration editing option, relative mV (RmV), ORP and temperature