Eco Best Technology® Nitrile Cleanroom Gloves, Powder-Free, Showa
Supplier: Showa-Best Glove
Certificates
About this item
Created with premium-grade plastizers and nitrile reinforcement, SHOWA 731 brings you the pinnacle of tactile precision and high-capacity protection against solvents and acids.
- 100% nitrile with Eco Best Technology®
- Unsupported, unlined
- 15 mil thick
- Bisque grip
These nitrile gloves provide chemical protection against solvents and acids, and their bisque finish grip makes wet work easier and safer. Effective gripping allows superior tactile precision.
Specifications
- Catalog No:
- 75804-786
- 75804-788
- 75804-790
- 75804-794
- 75804-784
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free