To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:In methylene chloride
- Description:Grob DA 280 Column Test Mix
- Size:1 ml/Ampule
- Cat. no.:75835-376
- Supplier No.:35252
Specifications
About this item
Contains: n-decane, C10, 280 µg/ml, methyl decanoate, C10:0, 420 µg/ml, n-undecane, C11, 280 µg/ml, methyl undecanoate, C11:0, 420 µg/ml, methyl dodecanoate, C12:0, 420 µg/ml, L(+)-2,3-butanediol, 530 µg/ml, 2,6-dimethylaniline, 320 µg/ml, 2,6-dimethylphenol, 320 µg/ml, 2-ethylhexanoic acid, 380 µg/ml, nonanal, 400 µg/ml, 1-octanol, 360 µg/ml.