Specifications
- Concentration:In methylene chloride:methanol (1:1)
- Description:Amine Column Test Mix
- Size:1 ml/Ampule
- Cat. no.:75835-326
- Supplier No.:35002
Specifications
About this item
The amine column test mix consists of (C10) decane (0,60 mg/ml), (C12) dodecane (0.60 mg/ml, 1,2-butanediol (0.60 mg/ml), diethanolamine (DEA) (1.20 mg/ml), diethylenetriamine (1.20 mg/ml), 2,6-dimethylaniline (0.60 mg/ml), 2-nonanol (0.60 mg/ml), pyridine(0.60 mg/ml).