To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:E1387 Column Resolution Check Mix
- Size:1 ml/Ampule
- Cat. no.:75834-636
- Supplier No.:31224
Specifications
About this item
This mix also can be used for underground storage tank monitoring.
It consist of 13 components: (C6) n-Hexane, (C8) n-Octane, (C10) n-Decane, (C12) n-Dodecane, (C14) n-Tetradecane, (C16) n-Hexadecane, (C18) n-Octadecane, (C20) n-Eicosane, 2-Ethyltoluene, 3-Ethyltoluene, Toluene, 1,2,4-Trimethylbenzene and p-Xylene.
Shelf life maximum 85 monthsa and minimum 6 months.