To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:100 µg/ml each in toluene
- Description:GC Multiresidue Pesticide Standard #1
- Size:1 ml/Ampule
- Cat. no.:75835-046
- Supplier No.:32563
Specifications
About this item
Organophosphorus compounds that has accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
Contains: Azinphos ethyl, azinphos methyl, chlorpyrifos, chlorpyrifos methyl, diazinon, EPN, fenitrothion, isazophos, phosalone, phosmet, pirimiphos ethyl, pirimiphos methyl, pyraclofos, pyrazophos, pyridaphenthion, quinalphos.