Specifications
- Concentration:300 µg/ml each in acetonitrile:acetic acid (99.9:0.1)
- Description:QuEChERS Performance Standard A
- Size:1 ml/Ampule
- Cat. no.:75834-626
- Supplier No.:31153
Specifications
About this item
QuEChERS performance standard A designed for use in all QuEChERS methods for pesticides in fruits and vegetables. Ideal for initial method evaluations and ongoing method performance validations.
- Optimized blend of chemically compatible analytes for maximum stability and shelf life
- Quantitatively analyzed to confirm the composition and stability of each mixture
Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards.
This standard contain 16 components: Acephate, Azinphos methyl, Chlorpyrifos, Coumaphos, Diazinon, Dichlofluanid, Dichlorvos, Dimethoate, Fenthion, Malathion, Methamidophos, Mevinphos, Omethoate, Phosalone, Pirimiphos methyl and Propargite.