To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:200 µg/ml each in acetonitrile
- Description:Steroids and Mixed Pharmaceuticals Mix
- Size:1 ml/Ampule
- Cat. no.:75834-622
- Supplier No.:31117
Specifications
About this item
This steroids and mixed pharmaceuticals mix contain ten components: Bisphenol A, Diclofenac sodium salt, 17-beta-Estradiol, Estrone, 17-alpha-Ethynylestradiol, 4-para-Nonylphenol, 4-tert-Octylphenol, Primidone, Progestrone and Testosterone.
Shelf life 24 months.