Specifications
- Concentration:100 mg/dl each in water
- Description:BAC Resolution Control Standard t-B
- Size:1 ml/Ampule
- Cat. no.:75835-434
- Supplier No.:36011
Specifications
About this item
The mix is used to verify the retention time for each compound normally included in a blood alcohol test, and to verify that the compounds are resolved from and do not interfere with one another.
- Includes tert-butanol internal standard
- Intended for qualitative use only
It consists of acetaldehyde, acetone, tert-butanol (TBA), ethanol (BAC), methanol and 2-propanol (isopropanol).