Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:NJDEP EPH 10/08 Rev. 2 Aromatics Calibration Standard
- Size:1 ml/Ampule
- Cat. no.:75834-478
- Supplier No.:30541
Specifications
About this item
Consists of 18 components, those are acenaphthene, acenaphthylene, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, chrysene, dibenzanthracene, fluoranthene, fluorene, indenopyrene, 2-methylnaphthalene, naphthalene, phenanthrene, 1,2,3-trimethylbenzene and pyrene
18 components