Specifications
- Concentration:1000 µg/ml each in methylene chloride
- Description:CLP 04.1 B/N MegaMix, Revision 2
- Size:1 ml/Ampule
- Cat. no.:75834-878
- Supplier No.:31903
Specifications
About this item
Consists of 47 components, those are acenaphthene, acenaphthylene, acetophenone, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, benzyl butyl phthalate, biphenyl, bismethane, bisether, bisphthalate, 4-bromophenyl phenyl ether, carbazole, 4-chloroaniline, 2-chloronaphthalene, 4-chlorophenyl phenyl ether, chrysene, dibenzanthracene, dibenzofuran, diethylphthalate, dimethylphthalate, di-n-butyl phthalate, 2,4-dinitrotoluene, 2,6-dinitrotoluene, di-n-octyl phthalate, diphenylamine *,fluoranthene, fluorene, hexachlorobenzene, hexachloro-1,3-butadiene , hexachlorocyclopentadiene, hexachloroethane, indenopyrene, isophorone, 2-methylnaphthalene, naphthalene, 2-nitroaniline, 3-nitroaniline, 4-nitroaniline, nitrobenzene, n-nitroso-di-n-propylamine, 2,2'-oxybis, phenanthrene and pyrene.