Specifications
- Concentration:50 µg/ml each in isooctane:ethyl acetate (4:1)
- Description:PBDE Mix
- Size:1 ml/Ampule
- Cat. no.:75835-158
- Supplier No.:33098
Specifications
About this item
The PBDE mix consists of five components, they are 2,2',4,4',5,5'-hexabromobiphenyl, 2,2',4,4',5,5'-Hexabromodiphenyl ether (BDE-153), 2,2',4,4',5-Pentabromodiphenyl ether (BDE-99) , 2,2',4,4',6-Pentabromodiphenyl ether (BDE-100) and 2,2',4,4'-Tetrabromodiphenyl ether (BDE-47).