Specifications
- Concentration:In methyl tert-butyl ether
- Description:515.4 Methylated Chlorinated Acids Mix
- Size:1 ml/Ampule
- Cat. no.:75835-876
- Supplier No.:32444
Specifications
About this item
Mix contains Acifluorfen methyl ester, Bentazon methyl ester, Chloramben methyl ester, Dalapon methyl ester, 2,4-D methyl ester, 2,4-DB methyl ester, DCPA methyl ester (Chlorthal-dimethyl), Dicamba methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Dichlorprop methyl ester, Dinoseb methyl ether, Pentachloroanisole, Picloram methyl ester, Quinclorac methyl ester, 2,4,5-T methyl ester, 2,4,5-TP.