Specifications
- Concentration:In water:dimethyl sulfoxide (80:20)
- Description:European Pharmacopoeia/ICH Class 1 Mix (revised)
- Size:1 ml/Ampule
- Cat. no.:75835-450
- Supplier No.:36261
Specifications
About this item
The european pharmacopoeia/ICH Class 1 mix consists of five components, they are benzene (2 μg/ml), carbon tetrachloride (4 μg/ml), 1,2-dichloroethane (5 μg/ml), 1,1-dichloroethene (8 μg/ml) and 1,1,1-trichloroethane (10 μg/ml).