Specifications
- Concentration:In acetone
- Description:Organophosphorus Pesticide Mix, European Formulation
- Size:1 ml/Ampule
- Cat. no.:75834-966
- Supplier No.:32418
Specifications
About this item
Contains 16 components, those are acephate, azinphos methyl, chlorpyrifos, demeton-s-methyl, dichlorvos, dimethoate, ethion, malathion, methamidophos, methidathion, omethoate, pirimiphos methyl, profenofos, prothiofos, pyrazophos and tolclofos-methyl.