To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:10 µg/ml each in isooctane
- Description:PCB Congener Standard #2
- Size:1 ml/Ampule
- Cat. no.:75834-940
- Supplier No.:32294
Specifications
About this item
Contains: 2,4,4'-trichlorobiphenyl (BZ #28), 2,2',5,5'-tetrachlorobiphenyl (BZ #52), 2,2',4,5,5'-pentachlorobiphenyl (BZ #101), 2,3',4,4',5-pentachlorobiphenyl (BZ #118), 2,2',3,4,4',5'-hexachlorobiphenyl (BZ #138), 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153), 2,2',3,4,4',5,5'-heptachlorobiphenyl (BZ #180).