Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:OLC 03.2 SVOA Deuterated Monitoring Compounds (DMC)
- Size:1 ml/Ampule
- Cat. no.:75834-772
- Supplier No.:31810
Specifications
About this item
This OLC 03.2 is an analytical method for the detection of low-concentration volatile, semivolatile, and pesticide/Aroclor organics in aqueous samples using deuterated monitoring compounds (DMCs) as sample-by-sample accuracy indicators.
It contains 16 components include: Acenaphthylene-d8, Anthracene-d10, Benzo(a)pyrene-d12, Bis-(2-chloroethyl)ether-d8, 4-Chloroaniline-d4, 2-Chlorophenol-d4, 2,4-Dichlorophenol-d3, Dimethylphthalate-d6, 4,6-Dinitro-2-methylphenol-d2, Fluorene-d10, 4-Methylphenol-d8, Nitrobenzene-d5, 2-Nitrophenol-d4, 4-Nitrophenol-d4, Phenol-d5 and Pyrene-d10.