Specifications
- Concentration:In methylene chloride:methanol (1:1)
- Description:610 PAH Calibration Mix B
- Size:1 ml/Ampule
- Cat. no.:75834-700
- Supplier No.:31455
Specifications
About this item
This 610 PAH calibration mix B idela for LC-UV detection.
It consists of 16 components: Acenaphthene (1000 μg/ml), Acenaphthylene (2000 μg/ml), Anthracene(100 μg/ml), Benz(a)anthracene (100 μg/ml), Benzo(a)pyrene (100 μg/ml), Benzo(b)fluoranthene (200 μg/ml), Benzo(k)fluoranthene (100 μg/ml), Benzo(ghi)perylene (200 μg/ml), Chrysene (100 μg/ml), Dibenz(a,h)anthracene (200 μg/ml), Fluoranthene (200 μg/ml), Fluorene (200 μg/ml), Indeno(1,2,3-cd)pyrene (100 μg/ml), Naphthalene (1000 μg/ml), Phenanthrene (100 μg/ml) and Pyrene (100 μg/ml).