Haloacetic Acid Methyl Ester Mix #1 (6 Components), Restek
Catalog #: 75835-740
Supplier: Restek
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1000 µg/ml each in methyl tert-butyl ether
- Description:Haloacetic Acid Methyl Ester Mix #1
- Size:1 ml/Ampule
- Cat. no.:75835-740
- Supplier No.:31645
Specifications
About this item
The mix consists of methyl bromochloroacetate, methyl dibromoacetate, methyl dichloroacetate, methyl monobromoacetate, methyl monochloroacetate, methyl trichloroacetate.
- Prepared from purified neat reference materials for the highest quality
- Lot-to-lot consistency ensures analytical reproducibility
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements. Meet Stage 1 requirements for US EPA Methods 552, 552.1 and 552.2 and meet requirements for Stage 2 DBPR.
Customers Who Bought This Also Bought