Specifications
- Concentration:500 µg/ml each in ethyl acetate
- Description:508.1 Calibration Mix #1
- Size:1 ml/Ampule
- Cat. no.:75834-222
- Supplier No.:32094
Specifications
About this item
Consists of 17 components, those are aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc , 4,4'-ddd, 4,4'-dde, 4,4'-ddt, dieldrin, endosulfan i, endosulfan ii, endosulfan sulfate, endrin, endrin aldehyde, heptachlor, heptachlor epoxide and methoxychlor.
17 components