To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:In methylene chloride 0.5 ml/ampule
- Description:Marine Oil Test Mix
- Size:0.5 ml
- Cat. no.:75835-374
- Supplier No.:35249
Specifications
About this item
Marine oil test mix consist of 20 components.
Contains: Methyl myristate, C14:0, 600 µg/ml, methyl myristoleate, C14:1, 100 µg/ml, methyl palmitate, C16:0, 1600 µg/ml, methyl palmitoleate, C16:1, 500 µg/ml, methyl stearate, C18:0, 500 µg/ml, methyl stearate, C18:0, 800 µg/ml, methyl oleate, C18:1, 1300 µg/ml, methyl vaccenate, C18:1, 400 µg/ml, methyl linoleate, C18:2, 200 µg/ml, methyl linolenate, C18:3, 200 µg/ml, methyl arachidate, C20:0, 100 µg/ml, methyl 11-eicosenoate, C20:1, 900 µg/ml, methyl 11-14-eicosadienoate, C20:2, 100 µg/ml, methyl arachidonate, C20:4, 300 µg/ml, methyl 11-14-17eicosatrienoate, C20:3, 100 µg/ml, methyl eicosapenataenoate, C20:5, 1000 µg/ml, methyl behenate, C22:0, 100 µg/ml, methyl erucate, C22:1, 300 µg/ml, methyl docosahexaenoate, C22:6, 1200 µg/ml, methyl lignocerate, C24:0, 100 µg/ml, methyl nervonate, C24:1, 100 µg/ml.