Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Description:CA WIP VOA Standard
- Cat. no.:75835-546
- Supplier No.:30236
Specifications
About this item
For monitoring samples suspected of gasoline contamination requires laboratories to calibrate and report these compounds.
It consists of benzene, chlorobenzene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, ethylbenzene, methyl tert-butyl ether, toluene, m-xylene, o-xylene, p-xylene.