Specifications
- Concentration:2000 µg/ml each in methylene chloride
- Description:DRO/ORO Range Calibration Standard
- Size:1 ml/Ampule
- Cat. no.:75834-802
- Supplier No.:31832
Specifications
About this item
This consists of n-decane (C10), n-dodecane (C12), n-tetradecane (C14), n-hexadecane (C16), n-octadecane (C18), n-eicosane (C20) ,n-docosane (C22) , n-tetracosane (C24), n-hexacosane (C26), n-octacosane (C28), n-triacontane (C30) and n-dotriacontane (C32).