To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Packaging:1 mL/ampul
- Description:Alaska UST Method AK101AA, Ver. 3-1-99
- Size:1 ml
- Supplier No.:30461
Specifications
About this item
Alaska Department of Environmental Conservation (ADEC) regulations indicate which products and indicator compounds are to be tested for each petroleum range
The analyst must use the following Alaska Series Methods or appropriate SW-846 Method for the indicator compounds. The Alaska UST procedurals manual indicates which products are to be tested for each petroleum range.