Specifications
- Concentration:50000 µg/ml in methylene chloride
- Description:Creosote oil
- Size:1 ml/Ampule
- Cat. no.:75834-812
- Supplier No.:31838
Specifications
About this item
This is for total petroleum hydrocarbon pattern recognition of creosote oil.
Creosote oil, a widely used wood preservative produced by distilling coal tar, contains chemicals that are classified as carcinogens (benzo(a)pyrene).