SP Wilmad-LabGlass Precision PTFE Stirrer Bearings, SP Industries
Supplier: SP Industries
Certificates
About this item
Uniquely designed and lubricant free, these precision stirrer/stirring bearings are self-aligning
Complete 10 mm diameter assembly includes PTFE stirrer bearing, PTFE bushing, saddle O-ring (LG-9470-300), threaded heavy wall borosilicate glass adapter, and top cap (LG-10818-114).
The saddle O-ring provides constant force and a positive seal on the stirring rod that allows vacuum operation to as low as 1 mm. Ground finish rods from other manufacturers are not recommended for use with this bearing.
Ordering information: See LG-9501 for polished stirrer rods or LG-9502 for stainless steel rods.
Customers Who Bought This Also Bought