SP Wilmad-LabGlass 3 Neck O-Ring Flange Reaction Vessel Covers for Size 359 O-Rings, SP Industries
Supplier: SP Industries
Certificates
About this item
Featuring a center neck and two side necks with full length outer Standard Taper joints, these borosilicate glass 3-neck reaction vessel lids/covers have an 6-5/8" flange O.D and come with a size 359 O-ring.
Designed to be used with 2000mL, 3000mL, and 4000mL cylindrical reaction vessels (LG-8075, LG-8079A, LG8079B, and LG-8079C).
Ordering information: Use LG-7316-108 clamps with these lids.
Customers Who Bought This Also Bought