Specifications
- Description:BBL™ Simmons Citrate Agar, Slants
- Size:10/sp
- Cat. no.:CA90001-196
- Supplier No.:221026
Specifications
About this item
Simmons Citrate Agar is a culture medium for the differentiation of gram-negative bacteria on the basis of citrate utilization.
- Nitrogen and carbon from sodium citrate and ammonium dihydrogen phosphate
- Organisms produce an alkaline reaction
- Bromthymol blue indicator shifts from green to blue with reaction
A positive reaction is indicated by growth with an intense blue color in the slant. A negative reaction is evidenced by no color change or traceable growth.