Pressure Regulator Fixtures, L774VR, WaterSaver Faucet
Catalog #: 97014-330
Supplier: WaterSaver Faucet
Specifications
- Description:Pressure Regulator, L774VR
- Cat. no.:97014-330
- Inlet Pressure:5–300 psi
- Outlet Pressure:5 - 125 psig
- Supplier No.:L774V-R
Specifications
About this item
Point-of-use regulation of high purity gases for installation on fume hoods. Used for compressed air and inert high purity gases, including nitrogen, helium, argon and oxygen.
Pressure regulator / gauge assembly is surface mounted on front face of fume hood
Non-relieving type with brass body and neoprene diaphragm. Regulator assembly is furnished with 1/4" IPS mounting shanks, locknuts and washers. Fixture is specially cleaned, lubricated, assembled and packaged for use with high purity gases, including oxygen.
Regulator assembly, valve and outlet fitting are piped together behind face of hood (pipe and fittings are not supplied). Gauge is installed on right side of regulator.
Customers Who Bought This Also Bought