To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Sterile Pack - Double Wrapping
- Size:20/sp
- Cat. no.:CA90004-996
- Plate Diameter:60 mm
- Supplier No.:297759
Specifications
About this item
Trypticase™ Soy Agar with 5% Sheep Blood (TSA II) is used for the growth of fastidious organisms and for the visualization of hemolytic reactions.
- Casein and soy peptones provide nutrients for growth
- Sodium chloride maintains osmotic equilibrium
- Enriched agar base is the solidifying agent
- Ensures the integrity of the sample
- Prevents the dish from opening unintentionally
- Protects media from accidental cross contamination
- Improves stability during stacking and storage
The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
Trypticase Soy Agar with 5% Sheep Blood is extensively used for the recovery and cultivation of fastidious microbial species and for the determination of hemolytic reactions which are important differentiating characteristics for bacteria, especially Streptococcus species.