Parabolic Collision Monkey and Hunter Apparatus
Catalog #: 470220-606
Supplier: OSAW INDUSTRIAL PRODUCTS SE
Specifications
- Description:Parabolic Collision Monkey and Hunter Apparatus
- Cat. no.:470220-606
- Supplier No.:SA702
Specifications
About this item
This apparatus illustrates the effect of gravity on projectile motion.
- Minimal Setup
- Unit Also Works as Trajectory Apparatus
- Large, Bright Parts
- Table-Clamp Mounting Plate
- For Illustrating the Effect of Gravity on Projectile Motion
Using a tube equipped with a spring-loaded launching piston, students can aim at the “monkey”—a ball held in place by an electromagnet. When the projectile is fired, the electromagnetic circuit breaks. The trajectory is easily visible thanks to the large, bright parts. An angle mirror sight helps speed the aiming process.
This unit includes a table-clamp mounting plate and can be used as a trajectory apparatus.
Ordering information: This kit includes a launcher, electromagnetic free fall unit, two no. of projectile balls, angle mirror accessory, and instruction manual. A support stand and 1.5V battery is required.
Customers Who Bought This Also Bought