Specifications
- Description:Kanamycin
- Size:1 x 50 disks
- Cat. no.:75838-868
- Volume:1000 mcg
- Supplier No.:Z7191
Specifications
About this item
Differentiates anaerobic bacteria on the basis of susceptibility to kanamycin.
- Each cartridge contains 50 disks
- Spring-loaded for smooth and reliable dispensing
- Differentiates anaerobic bacteria
Differentiates anaerobic bacteria on the basis of susceptibility to kanamycin. Each cartridge contains 50 disks that are spring-loaded for smooth and reliable dispensing from a single metal disk dispenser.