Bottles, HDPE, Narrow Mouth, with Screw Cap
Catalog #: 470178-134
Supplier: Dynalon
Specifications
- Capacity:125 ml
- Color:Clear
- Cat. no.:470178-134
- Supplier No.:301715-0004
Specifications
About this item
Unbreakable and Leak-proof.
- Each bottle has screw cap with protective inner lining
- Use amber bottles for light sensitive liquids
These polyethylene bottles are resistant to most acids, bases, alcohols, aliphatic hydrocarbons, and ketones.
Customers Who Bought This Also Bought