Phosphate Buffer with Magnesium Chloride, Hardy Diagnostics
Catalog #: CA76006-308
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Packaging:Polyethylene terephthalate bottle
- Description:Phosphate buffer with magnesium chloride
- Cat. no.:CA76006-308
- Volume:1 L
Specifications
About this item
For the preparation of dilutions in plate counts for the dairy and food industries.
Customers Who Bought This Also Bought