BsrFI-v2, New England Biolabs
Catalog #: CA103218-954
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:BsrFI-v2
- Size:1000 units
- Cat. no.:CA103218-954
Specifications
About this item
A restriction endonuclease that recognizes the sequence R^CCGG_Y.
- Sourced from an E.coli strain that carries the cloned and modified BsrFI gene
- Cleavage of mammalian genomic DNA is blocked by CpG methylation
Customers Who Bought This Also Bought