a1-3,4 Fucosidase, New England Biolabs
Catalog #: CA102969-118
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:a1-3,4 Fucosidase
- Size:200 Units
- Cat. no.:CA102969-118
Specifications
About this item
α1-3,4 Fucosidase, (also known as AMF) is a broad specificity exoglycosidase that catalyzes the hydrolysis of α1-3 and α1-4 linked fucose residues from oligosaccharides and glycoproteins.
Customers Who Bought This Also Bought