WorkSafe Grease Resistant Rubber Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
Certificates
About this item
A matting solution designed for cutting fluid environments, where sure footing and proper drainage are essential.
- Lightweight
- High performance
- Cutting fluid resistant
An ideal anti-fatigue mat for wet industrial areas, WorkSafe Light has a well-engineered drainage system and molded-in edges to provide safe access. Ideally suited for use in oily environments.
Certifications: Meets ASTM F1677. Dry: COF = > 0.90/Wet: COF = 0.36. Meets ASTM D575. Deflection @35 psi, mm=4.10.
Customers Who Bought This Also Bought