DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus), Calbiochem®, Millipore®
Catalog #: CA80051-358
Supplier: MilliporeSigma
CAS Number: 9007-49-2
Specifications
- Packaging:Glass bottle
- Size:1 g
- Cat. no.:CA80051-358
- Supplier No.:2618-1GM
Specifications
Specification Test Results
- hyperchromicity:≥27%
- S20w:15.4-17.4
- A260/280 nm:≥ 1.8
- Calbiochem®, Millipore®, DNA sodium salt (Deoxyribonucleic acid sodium salt) (from calf thymus):
Specification Test Results
About this item
Synonyms: Thymonucleic acid sodium salt
:Storage Temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. N/A N/A
Storage Temperature:Freezer
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130921
Customers Who Bought This Also Bought