About this item
The M1/69 antibody reacts with mouse CD24, a cell surface molecule also known as Heat Stable Antigen (HSA). CD24 is a GPI-linked glycoprotein involved in cell adhesion and is a ligand for CD62P (P-selectin). It is expressed on erthrocytes, thymocytes, peripheral lymphocytes and myeloid lineage cells. The expression of CD24 is used to delineate stages of of lymphocyte development in the bone marrow.
Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation: APC
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75834-302
- 75834-304
- Antigen name:
- CD24, Heat Stable Antigen
- CD24, Heat Stable Antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- M1/69
- M1/69
- Host:
- Rat
- Rat
- Gene ID:
- 12484
- 12484
- Isotype:
- IgG2b kappa
- IgG2b kappa