Specifications
- Description:Determination of the Alkalinity of Water
- Cat. no.:470146-440
- Supplier No.:IS9752
- No. of Uses:40
Specifications
About this item
This kit allows analysts to perform a very simple titration to determine the alkalinity and the phenolphthalein of their fresh water sample.
- Hands-on student activity
- Students can learn to titrate in the lab or in the field
- Activity explains water's ability to neutralize acids
- Lab provides instructions
- Solutions included
- Aligned with GLOBE protocols
The titration uses an easily identifiable color transition to indicate the end of the analysis.
The kit contains everything an analyst would need to perform a water analysis either in the laboratory setting or out in the field.