Hairnets, Cellucap-Melco
Catalog #: 10833-038
Supplier: Cellucap-Melco
Specifications
- Ø:20"
- Garment application:Food,Industrial,Laboratory
- Color:White
- Description:Hairnets, K100 Style
- Cat. no.:10833-038
- Supplier No.:K120FDWT
Specifications
About this item
Polyester honeycomb hairnets are ideal for use in food processing, safety and industrial applications
This single strand 30 denier net is made with a 1/8" aperture for better hair containment.
Customers Who Bought This Also Bought