ContactUsLinkComponent
HardyDisks™ AST XV Factor HardyDisks™, Hemin and NAD Disk, Hardy Diagnostics
HardyDisks™ AST XV Factor HardyDisks™, Hemin and NAD Disk, Hardy Diagnostics
Catalog #: 75832-734
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
XV Factor, HardyDisks, Hemin and NAD disk
HardyDisks™ AST XV Factor HardyDisks™, Hemin and NAD Disk, Hardy Diagnostics
Catalog #: 75832-734
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Packaging:
    1 x 50 disks
  • Description:
    XV Factor HardyDisks™, Hemin and NAD Disk
  • Cat. no.:
    75832-734
  • Supplier No.:
    Z7051
Specifications
About this item
XV Factor HardyDisks™, Hemin and NAD disk is designed to aid in the identification of Haemophilus spp.
Contains NAD and Hemin (X and V Factors) for the speciation of Haemophilus. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences. The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP), and have achieved ISO 13485 certification as a medical device manufacturer.
Customers Who Bought This Also Bought