2,2,4-Trimethylpentane ≥99.75% PRF, B&J Brand™ ASTM D2699/D2700, Burdick & Jackson™
Supplier: Burdick & Jackson
Certificates
About this item
Synonyms: Isobutyltrimethylmethane,Isooctane
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MW:114.23 g/mol
Boiling Pt:99.24 °C
Melting Pt:-107.4 °C
Density:0.69 g/cm³ (20 °C)
Flash Pt:-8 °C
MDL Number:MFCD00008943
CAS Number:540-84-1
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,75
:Boiling Pt:
:Boiling Pt: 99.24 °C
:Melting Pt:
:Melting Pt: -107.4 °C
:Flash Pt:
:Flash Pt: -8 °C
:M.W. 114.23 g/mol
EINECS:208-759-1
:99.24 °C
UN:1262
:3
:II
:-8 °C
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:Clear, colorless liquid
- Color (APHA):10
- Water (Karl Fischer):0.010%
- Residue:5 mg/L
- GC Purity:99.75%
- n-Heptane:0.1%
- Lead Content:0.5 mg/L
- Sulfur Compounds (as S):0.005%
- Burdick & Jackson™, 2,2,4-Trimethylpentane ≥99.75% PRF, B&J Brand™ ASTM D2699/D2700:
Specification Test Results
Related Information
Certificates
Search by Lot NumberHoneywell Research Chemicals Primary Reference Fuels Flyer
Product Literature
TDS Comparing the Leading Solvents
Datasheet
Customers Who Bought This Also Bought