Specifications
- Description:Cut Down Tray
- Cat. no.:75817-524
- Packaging:10/Case
Specifications
About this item
Cut Down Trays are industry standard
Each tray includes the following instruments and prep materials: #11 Long Scalpel, #15 Long Scalpel, OR Scissors, Straight Hemostat, Dressing Forceps, (2) Straight Mosquito Forceps, Hemostats, (3) Curved Mosquito Hemostats, Adson Tissue Forceps, Mayo-Hegar Needle Holder, Webster Needle Holder, (2) Blue Linen OR Towels, (15) Gauze Sponges (4x4", 12-ply), (2) Instrument Trays, and Clear Utility Drape.