Plankton Slide
Catalog #: 470176-932
Supplier: Avantor
Specifications
- Description:Plankton
- Cat. no.:470176-932
- Stain:Fast Green and Mallory's
- Supplier No.:WARD470176-932
- Orientation:Whole mount
Specifications
About this item
A Mixture of Small Organisms and Diatoms
- Identifying features clearly distinguishable
- Rigorous quality control standards
Shows general structures.
Customers Who Bought This Also Bought