About this item
Versatile and reliable. The Innova 42 and 42R (refrigerated) incubator shakers are stackable, can be placed on or under the bench, and come with a static shelf to be used to incubate Agar plates while shaking cultures in flasks.
- High capacity in a small footprint: for example, 4×5 or 2×6 L flasks on universal platform; device footprint 4731 cm²/733 in² (63.5 cm/25" width, 74.5 cm/29.3" depth)
- Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes and can be reached easily even in a double stack
- Easy to clean: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
- Lab position flexibility: Lab position on or under bench, 2× stackable
- Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
- Sample evaporation protection: Bottom spill pan usable as water reservoir; optional culture drawer, for Agar plates, for example
- Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
- Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
- Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
The Innova 42 and 42R (refrigerated) shakers are very versatile incubator shakers in terms of placement and options. They can be stacked up to two units high and can also be placed on or under a standard height laboratory bench. In every scenario, the side opening door/s allow users access right up to the chamber. Options include a static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan. Additionally, a photosynthetic light bank and UV germicidal light are available for the 42R.
The cast iron triple-eccentric counterbalanced drive is specifically designed to support high-speed applications and heavy workloads - the design has stood the test of time for over 70 years and continues to enable scientists today. The simple user interface allows for easy programming of speed, temperature, time and when installed, the photosynthetic light bank. The VisioNize box can be added to enhance monitoring and adds notifications functionality.
Specifications
- Catalog No:
- CA75875-642
- CA75875-640
- CA75875-644
- CA75875-638
- Temperature range:
- 42: 10 °C above ambient to 80 °C
42R: 20 °C below ambient to 80 °C (min. 4 °C) - 42: 10 °C above ambient to 80 °C
42R: 20 °C below ambient to 80 °C (min. 4 °C) - 42: 10 °C above ambient to 80 °C
42R: 20 °C below ambient to 80 °C (min. 4 °C) - 42: 10 °C above ambient to 80 °C
42R: 20 °C below ambient to 80 °C (min. 4 °C)
- W×D×H:
- 63.5×74.5×81.8 cm (25×29.3×32.8")
- 63.5×74.5×81.8 cm (25×29.3×32.8")
- 63.5×74.5×81.8 cm (25×29.3×32.8")
- 63.5×74.5×81.8 cm (25×29.3×32.8")
- Speed range:
- 25 - 400 rpm; 25 - 300 rpm when using flasks >500 ml in stacked devices
- 25 - 400 rpm; 25 - 300 rpm when using flasks >500 ml in stacked devices
- 25 - 400 rpm; 25 - 300 rpm when using flasks >500 ml in stacked devices
- 25 - 400 rpm; 25 - 300 rpm when using flasks >500 ml in stacked devices
- Outside depth:
- 131 cm (51.6") with door open
- 131 cm (51.6") with door open
- 131 cm (51.6") with door open
- 131 cm (51.6") with door open
- Memory:
- Non volatile with automatic power failure restart
- Non volatile with automatic power failure restart
- Non volatile with automatic power failure restart
- Non volatile with automatic power failure restart
- Format:
- Benchtop, under bench, stackable (×2)
- Benchtop, under bench, stackable (×2)
- Benchtop, under bench, stackable (×2)
- Benchtop, under bench, stackable (×2)
- Electrical:
- 120 V, 60 Hz
- 120 V, 60 Hz
- 120 V, 60 Hz
- 120 V, 60 Hz